Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-10 Kit
SDP
Catalog No. 5000079051
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IL-10 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 10 (IL-10) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluent The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for cell culture supernatants The total assay time is 3 5 hours This kit can be used to measure mouse IL-10 levels for immunology and inflammation research
Catalog No. 50-000-79051 Supplier Meso Scale Discovery Supplier No. K152QUG1
The Meso Scale Discovery V-PLEX Plus Mouse IL-10 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 10 (IL-10) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluent The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for cell culture supernatants The total assay time is 3 5 hours This kit can be used to measure mouse IL-10 levels for immunology and inflammation research