Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-12p70 Kit
SDP
Catalog No. 5000078309
Encompass
The V-PLEX Plus Mouse IL-12p70 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-12p70 in mouse serum plasma and cell culture supernatants The kit includes a plate coated with a mouse IL-12p70 capture antibody a ruthenium-labeled mouse IL-12p70 detection antibody calibrators and buffers for sample dilution The assay has a sensitivity of 1 3 pg/mL and a broad dynamic range from 1 3-1000 pg/mL It requires a small sample volume of 25 L and the total assay time is 3 5 hours This ECL immunoassay allows for the sensitive and accurate quantification of IL-12p70 levels in mouse biological samples
Catalog No. 50-000-78309 Supplier Meso Scale Discovery Supplier No. K152QVG1
The V-PLEX Plus Mouse IL-12p70 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-12p70 in mouse serum plasma and cell culture supernatants The kit includes a plate coated with a mouse IL-12p70 capture antibody a ruthenium-labeled mouse IL-12p70 detection antibody calibrators and buffers for sample dilution The assay has a sensitivity of 1 3 pg/mL and a broad dynamic range from 1 3-1000 pg/mL It requires a small sample volume of 25 L and the total assay time is 3 5 hours This ECL immunoassay allows for the sensitive and accurate quantification of IL-12p70 levels in mouse biological samples