Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-12p70 Kit
SDP
Catalog No. 5000078440
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IL-12p70 Kit is an electrochemiluminescence immunoassay for the quantitative measurement of mouse interleukin 12 subunit p70 (IL-12p70) in cell culture supernatants serum plasma and other biological fluids This kit contains a plate coated with a mouse IL-12p70 capture antibody a ruthenium-labeled mouse IL-12p70 detection antibody calibrators and buffers for a fast and sensitive chemiluminescent immunoassay with a minimum detectable dose of 0 26 pg/mL IL-12p70 The kit provides components for one 96-well plate allowing for the quantitative measurement of mouse IL-12p70 in biological samples
Catalog No. 50-000-78440 Supplier Meso Scale Discovery Supplier No. K152QVG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Mouse IL-12p70 Kit is an electrochemiluminescence immunoassay for the quantitative measurement of mouse interleukin 12 subunit p70 (IL-12p70) in cell culture supernatants serum plasma and other biological fluids This kit contains a plate coated with a mouse IL-12p70 capture antibody a ruthenium-labeled mouse IL-12p70 detection antibody calibrators and buffers for a fast and sensitive chemiluminescent immunoassay with a minimum detectable dose of 0 26 pg/mL IL-12p70 The kit provides components for one 96-well plate allowing for the quantitative measurement of mouse IL-12p70 in biological samples