Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-15 Kit
SDP
Catalog No. 5000079096
Encompass
The Meso Scale Discovery Mouse IL-15 Kit is a sensitive immunoassay for detecting and quantifying mouse interleukin 15 (IL-15) in samples like serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for rapid accurate IL-15 quantification with high sensitivity and a broad range Detection limits are 1 3 pg/mL to 10 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent The assay takes about 3 5 hours
Catalog No. 50-000-79096 Supplier Meso Scale Discovery Supplier No. K152RDG2
Add to Cart
Add to Cart
The Meso Scale Discovery Mouse IL-15 Kit is a sensitive immunoassay for detecting and quantifying mouse interleukin 15 (IL-15) in samples like serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for rapid accurate IL-15 quantification with high sensitivity and a broad range Detection limits are 1 3 pg/mL to 10 000 pg/mL The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent The assay takes about 3 5 hours