Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-15 Kit
SDP
Catalog No. 5000079317
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 15 (IL-15) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 5-2500 pg/mL and takes approximately 3 5 hours to complete It uses an instrument such as the MSD QuickPlex SQ 120 for plate reading The kit has high sensitivity wide dynamic range and requires low sample volume
Catalog No. 50-000-79317 Supplier Meso Scale Discovery Supplier No. K152RDG1
The Meso Scale Discovery V-PLEX Plus Mouse IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 15 (IL-15) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 5-2500 pg/mL and takes approximately 3 5 hours to complete It uses an instrument such as the MSD QuickPlex SQ 120 for plate reading The kit has high sensitivity wide dynamic range and requires low sample volume