Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-15 Kit
SDP
Catalog No. 5000079328
Encompass
The Meso Scale Discovery Mouse IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 15 (IL-15) in biological samples such as serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay The kit can accurately quantify mouse IL-15 over a range of 3 2-10 000 pg/mL
Catalog No. 50-000-79328 Supplier Meso Scale Discovery Supplier No. K152RDG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Mouse IL-15 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 15 (IL-15) in biological samples such as serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay The kit can accurately quantify mouse IL-15 over a range of 3 2-10 000 pg/mL