Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-17A/F Kit
SDP
Catalog No. 5000079058
Encompass
The V-PLEX Mouse IL-17A/F Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of mouse interleukin 17A and 17F in serum plasma cell culture supernatants and other biological fluids The assay uses MSD s proprietary V-PLEX technology includes a plate coated with capture antibodies detection antibodies labeled with an ECL compound and calibrator solutions for generating a standard curve The kit provides high sensitivity requires low sample volume and has a rapid protocol that allows results in as little as 3 5 hours
Catalog No. 50-000-79058 Supplier Meso Scale Discovery Supplier No. K152WNG1
The V-PLEX Mouse IL-17A/F Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of mouse interleukin 17A and 17F in serum plasma cell culture supernatants and other biological fluids The assay uses MSD s proprietary V-PLEX technology includes a plate coated with capture antibodies detection antibodies labeled with an ECL compound and calibrator solutions for generating a standard curve The kit provides high sensitivity requires low sample volume and has a rapid protocol that allows results in as little as 3 5 hours