Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-17A/F Kit
SDP
Catalog No. 5000079059
Encompass
Meso Scale Diagnostics offers the V-PLEX Plus Cytokine Panel an electrochemiluminescent multiplex immunoassay for measuring multiple cytokines chemokines growth factors and immunoglobulins from one sample It uses patented MULTI-ARRAY technology with V-PLEX Plus plates for greater sensitivity and dynamic range The panel includes over 45 analytes from various species with high specificity and sensitivity for inflammation immunity and angiogenesis It requires low sample volume and provides high-throughput results with quantitative data
Catalog No. 50-000-79059 Supplier Meso Scale Discovery Supplier No. K152WNG2
Meso Scale Diagnostics offers the V-PLEX Plus Cytokine Panel an electrochemiluminescent multiplex immunoassay for measuring multiple cytokines chemokines growth factors and immunoglobulins from one sample It uses patented MULTI-ARRAY technology with V-PLEX Plus plates for greater sensitivity and dynamic range The panel includes over 45 analytes from various species with high specificity and sensitivity for inflammation immunity and angiogenesis It requires low sample volume and provides high-throughput results with quantitative data