Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-17A/F Kit
SDP
Catalog No. 5000080338
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-17A/F Kit is a multiplex immunoassay for measuring mouse interleukin 17A (IL-17A) and interleukin 17F (IL-17F) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection with a broad dynamic range The assay requires only 50 L of sample and has a 2 5-hour protocol The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrator blend and assay diluent enabling efficient quantification of IL-17A and IL-17F
Catalog No. 50-000-80338 Supplier Meso Scale Discovery Supplier No. K152WNG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Mouse IL-17A/F Kit is a multiplex immunoassay for measuring mouse interleukin 17A (IL-17A) and interleukin 17F (IL-17F) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection with a broad dynamic range The assay requires only 50 L of sample and has a 2 5-hour protocol The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrator blend and assay diluent enabling efficient quantification of IL-17A and IL-17F