Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-1SY Kit
SDP
Catalog No. 5000078443
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-1 Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 1 beta (IL-1 ) in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluents wash buffers read buffer and adhesive seals The detection range is 0 06-1500 pg/mL with a 25 L sample volume It uses the MSD SECTOR Imager offering high sensitivity and a wide dynamic range without sample dilution The kit lasts 9 months at 2-8 C
Catalog No. 50-000-78443 Supplier Meso Scale Discovery Supplier No. K152QPG2
The Meso Scale Discovery V-PLEX Mouse IL-1 Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 1 beta (IL-1 ) in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluents wash buffers read buffer and adhesive seals The detection range is 0 06-1500 pg/mL with a 25 L sample volume It uses the MSD SECTOR Imager offering high sensitivity and a wide dynamic range without sample dilution The kit lasts 9 months at 2-8 C