Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-1SY Kit
SDP
Catalog No. 5000078711
Encompass
The V-PLEX Plus Mouse IL-1 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of mouse interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and tissue lysates The assay uses a plate that has been pre-coated with capture antibodies specific for mouse IL-1 Samples and standards are added to the wells followed by the addition of a detection antibody conjugated to an ECL label Levels of mouse IL-1 are determined using an MSD instrument to measure ECL signals The kit includes a plate detection antibody calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator
Catalog No. 50-000-78711 Supplier Meso Scale Discovery Supplier No. K152QPG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Mouse IL-1 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of mouse interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and tissue lysates The assay uses a plate that has been pre-coated with capture antibodies specific for mouse IL-1 Samples and standards are added to the wells followed by the addition of a detection antibody conjugated to an ECL label Levels of mouse IL-1 are determined using an MSD instrument to measure ECL signals The kit includes a plate detection antibody calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator