Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-1SY Kit
SDP
Catalog No. 5000081512
Encompass
The V-PLEX Plus Mouse IL-1 Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) to measure mouse interleukin 1 beta (IL-1 ) in various samples It employs a V-PLEX sandwich format with antibodies on carbon electrodes in 96-well plates Samples and standards are added followed by an ECL-labeled detection antibody Light emission is quantified upon electricity application The kit includes a plate detection antibody calibrator and diluent offering high sensitivity and a dynamic range of 1 5 to 10 000 pg/mL
Catalog No. 50-000-81512 Supplier Meso Scale Discovery Supplier No. K152QPG1
The V-PLEX Plus Mouse IL-1 Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) to measure mouse interleukin 1 beta (IL-1 ) in various samples It employs a V-PLEX sandwich format with antibodies on carbon electrodes in 96-well plates Samples and standards are added followed by an ECL-labeled detection antibody Light emission is quantified upon electricity application The kit includes a plate detection antibody calibrator and diluent offering high sensitivity and a dynamic range of 1 5 to 10 000 pg/mL