Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-2 Kit
SDP
Catalog No. 5000078310
Encompass
Meso Scale Diagnostics offers the V-PLEX Plus Cytokine Panel an electrochemiluminescent multiplex immunoassay for measuring multiple cytokines chemokines growth factors and immunoglobulins from a single sample This sensitive assay measures biomarkers from human serum plasma and culture supernatants providing quantitative results in 3 5 hours with just 25-50 L of sample It supports research in immunology oncology neurobiology and more with various analyte combinations enabling efficient immune monitoring
Catalog No. 50-000-78310 Supplier Meso Scale Discovery Supplier No. K152QQG4
Add to Cart
Add to Cart
Request a Quote
Meso Scale Diagnostics offers the V-PLEX Plus Cytokine Panel an electrochemiluminescent multiplex immunoassay for measuring multiple cytokines chemokines growth factors and immunoglobulins from a single sample This sensitive assay measures biomarkers from human serum plasma and culture supernatants providing quantitative results in 3 5 hours with just 25-50 L of sample It supports research in immunology oncology neurobiology and more with various analyte combinations enabling efficient immune monitoring