Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-2 Kit
SDP
Catalog No. 5000078648
Encompass
The Meso Scale Discovery IL-2 Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for quantifying human interleukin-2 (IL-2) in serum plasma tissue culture media and other samples It detects 0 07-2500 pg/mL using just 25 L of sample The kit employs MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity wide range and fast results It includes a 96-well plate with IL-2 capture antibody detection antibody with an electrochemiluminescent label calibrators diluent read buffer and plate seals The assay completes in about 3 5 hours
Catalog No. 50-000-78648 Supplier Meso Scale Discovery Supplier No. K152QQG2
The Meso Scale Discovery IL-2 Ultra-Sensitive Kit is an electrochemiluminescent immunoassay for quantifying human interleukin-2 (IL-2) in serum plasma tissue culture media and other samples It detects 0 07-2500 pg/mL using just 25 L of sample The kit employs MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity wide range and fast results It includes a 96-well plate with IL-2 capture antibody detection antibody with an electrochemiluminescent label calibrators diluent read buffer and plate seals The assay completes in about 3 5 hours