Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-2 Kit
SDP
Catalog No. 5000079362
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 2 (IL-2) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibodies against mouse IL-2 detection antibodies labeled with an electrochemiluminescent compound and calibrator containing recombinant mouse IL-2 The assay has a detection range from 3 2 to 10 000 pg/mL and exhibits no cross-reactivity with other cytokines This highly sensitive and reproducible kit allows rapid quantification of mouse IL-2 levels
Catalog No. 50-000-79362 Supplier Meso Scale Discovery Supplier No. K152QQG1
The Meso Scale Discovery V-PLEX Plus Mouse IL-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 2 (IL-2) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibodies against mouse IL-2 detection antibodies labeled with an electrochemiluminescent compound and calibrator containing recombinant mouse IL-2 The assay has a detection range from 3 2 to 10 000 pg/mL and exhibits no cross-reactivity with other cytokines This highly sensitive and reproducible kit allows rapid quantification of mouse IL-2 levels