Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-33 Kit
SDP
Catalog No. 5000079062
Encompass
The K152XBG-4 is a 4-channel 16-bit analog output module from Keithley Instruments It provides programmable 10V analog outputs with 16-bit resolution The module has 4 isolated channels that can output bipolar voltages It is compatible with the KPCI-3108 Series data acquisition cards allowing for analog signal generation The module is hot swappable for easy installation and removal Key specifications include 10V output range 16-bit resolution 4 isolated channels and 0 012% FSR accuracy The K152XBG-4 is well-suited for precision analog signal generation in automated test and measurement applications
Catalog No. 50-000-79062 Supplier Meso Scale Discovery Supplier No. K152XBG4
Add to Cart
Add to Cart
Request a Quote
The K152XBG-4 is a 4-channel 16-bit analog output module from Keithley Instruments It provides programmable 10V analog outputs with 16-bit resolution The module has 4 isolated channels that can output bipolar voltages It is compatible with the KPCI-3108 Series data acquisition cards allowing for analog signal generation The module is hot swappable for easy installation and removal Key specifications include 10V output range 16-bit resolution 4 isolated channels and 0 012% FSR accuracy The K152XBG-4 is well-suited for precision analog signal generation in automated test and measurement applications