Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-33 Kit
SDP
Catalog No. 5000082245
Encompass
The Meso Scale Discovery V-PLEX Human IL-33 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-33 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a detection range of 0 05-1000 pg/mL and a minimum detectable dose of 0 03 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium read buffer wash buffer blocker solutions calibrator diluent and a vial of lyophilized calibrator The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-82245 Supplier Meso Scale Discovery Supplier No. K152XBG2
The Meso Scale Discovery V-PLEX Human IL-33 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-33 in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a detection range of 0 05-1000 pg/mL and a minimum detectable dose of 0 03 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium read buffer wash buffer blocker solutions calibrator diluent and a vial of lyophilized calibrator The assay takes approximately 3 5 hours to complete