Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-33 Kit
SDP
Catalog No. 5000082411
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IL-33 Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 33 (IL-33) in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibody detection antibody with electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay sensitivity is 19 5 pg/mL with a range of 78 1-5000 pg/mL requiring only 50 L of sample It can assay 46 samples in duplicate
Catalog No. 50-000-82411 Supplier Meso Scale Discovery Supplier No. K152XBG1
The Meso Scale Discovery V-PLEX Plus Mouse IL-33 Kit is an electrochemiluminescent immunoassay for quantifying mouse interleukin 33 (IL-33) in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibody detection antibody with electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay sensitivity is 19 5 pg/mL with a range of 78 1-5000 pg/mL requiring only 50 L of sample It can assay 46 samples in duplicate