Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-4 Kit
SDP
Catalog No. 5000079353
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 03-2500 pg/mL and takes approximately 3 5 hours to complete It uses an instrument such as the MESO QuickPlex SQ 120 for plate reading The kit can be used to measure natural and recombinant mouse IL-4
Catalog No. 50-000-79353 Supplier Meso Scale Discovery Supplier No. K152QRG1
The Meso Scale Discovery V-PLEX Plus Mouse IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 03-2500 pg/mL and takes approximately 3 5 hours to complete It uses an instrument such as the MESO QuickPlex SQ 120 for plate reading The kit can be used to measure natural and recombinant mouse IL-4