Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-4 Kit
SDP
Catalog No. 5000080432
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue lysates The assay utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence detection to provide high sensitivity broad dynamic range and rapid results The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions to generate a standard curve The assay has high sensitivity with a lower limit of detection of 0 04 pg/mL and an upper limit of 10 000 pg/mL
Catalog No. 50-000-80432 Supplier Meso Scale Discovery Supplier No. K152QRG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Mouse IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue lysates The assay utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence detection to provide high sensitivity broad dynamic range and rapid results The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions to generate a standard curve The assay has high sensitivity with a lower limit of detection of 0 04 pg/mL and an upper limit of 10 000 pg/mL