Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-4 Kit
SDP
Catalog No. 5000082120
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue lysates The assay utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence detection to provide high sensitivity broad dynamic range and rapid results The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions to generate a standard curve The assay has a detection range from 0 03-2500 pg/mL and requires 50 L or less sample volume per well
Catalog No. 50-000-82120 Supplier Meso Scale Discovery Supplier No. K152QRG2
The Meso Scale Discovery V-PLEX Mouse IL-4 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue lysates The assay utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence detection to provide high sensitivity broad dynamic range and rapid results The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions to generate a standard curve The assay has a detection range from 0 03-2500 pg/mL and requires 50 L or less sample volume per well