Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-5 Kit
SDP
Catalog No. 5000079316
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 5 (IL-5) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and a sample volume requirement of 25 L The total assay time is approximately 3 5 hours This kit allows for the sensitive and accurate quantification of mouse IL-5
Catalog No. 50-000-79316 Supplier Meso Scale Discovery Supplier No. K152QSG1
The Meso Scale Discovery V-PLEX Plus Mouse IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 5 (IL-5) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and a sample volume requirement of 25 L The total assay time is approximately 3 5 hours This kit allows for the sensitive and accurate quantification of mouse IL-5