Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-6 Kit
SDP
Catalog No. 5000079358
Encompass
Meso Scale Diagnostics offers the V-PLEX Plus Cytokine Panel 1 (human) Kit which is used to simultaneously quantify multiple protein biomarkers in serum plasma and culture supernatants This electrochemiluminescence immunoassay allows researchers to accurately measure up to 10 cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) from a single small sample volume The kit provides high sensitivity a broad dynamic range and requires less hands-on time compared to traditional ELISA assays With a streamlined and optimized protocol the V-PLEX Plus Cytokine Panel 1 Kit enables efficient and reproducible analysis of cytokine signaling networks
Catalog No. 50-000-79358 Supplier Meso Scale Discovery Supplier No. K152QXG4
Add to Cart
Add to Cart
Request a Quote
Meso Scale Diagnostics offers the V-PLEX Plus Cytokine Panel 1 (human) Kit which is used to simultaneously quantify multiple protein biomarkers in serum plasma and culture supernatants This electrochemiluminescence immunoassay allows researchers to accurately measure up to 10 cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) from a single small sample volume The kit provides high sensitivity a broad dynamic range and requires less hands-on time compared to traditional ELISA assays With a streamlined and optimized protocol the V-PLEX Plus Cytokine Panel 1 Kit enables efficient and reproducible analysis of cytokine signaling networks