Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-6 Kit
SDP
Catalog No. 5000078305
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 6 (IL-6) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-1500 pg/mL and takes approximately 3 5 hours to complete It uses an instrument such as the MESO QuickPlex SQ 120 for plate reading The kit includes reagents for one 96-well plate and can assay 40 samples in duplicate
Catalog No. 50-000-78305 Supplier Meso Scale Discovery Supplier No. K152QXG1
The Meso Scale Discovery V-PLEX Plus Mouse IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 6 (IL-6) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-1500 pg/mL and takes approximately 3 5 hours to complete It uses an instrument such as the MESO QuickPlex SQ 120 for plate reading The kit includes reagents for one 96-well plate and can assay 40 samples in duplicate