Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-9 Kit
SDP
Catalog No. 5000079105
Encompass
The K152XCG-2 is a high-performance thermoelectric cooler from Mesoscale It has a maximum heat pumping capacity of 152W and can achieve a maximum temperature differential of 70 C The cooler has a footprint of 40 x 40 mm and incorporates a bonded semiconductor thermoelectric device between two ceramic plates to provide cooling or heating It requires a controlled DC power supply up to 15 2V for operation The K152XCG-2 can be used for a variety of precision thermal management applications
Catalog No. 50-000-79105 Supplier Meso Scale Discovery Supplier No. K152XCG2
The K152XCG-2 is a high-performance thermoelectric cooler from Mesoscale It has a maximum heat pumping capacity of 152W and can achieve a maximum temperature differential of 70 C The cooler has a footprint of 40 x 40 mm and incorporates a bonded semiconductor thermoelectric device between two ceramic plates to provide cooling or heating It requires a controlled DC power supply up to 15 2V for operation The K152XCG-2 can be used for a variety of precision thermal management applications