Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-9 Kit
SDP
Catalog No. 5000081996
Encompass
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit is a multiplex immunoassay measuring 30 human cytokines and chemokines in serum plasma and culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and adhesive seals The assay runs in 3 5 hours quantifying IL-1 IL-2 IL-4 IL-5 IL-6 IL-7 IL-8 IL-10 IL-12p70 IL-13 IL-15 IL-16 IL-17A TNF- IFN- GM-CSF VEGF-A CCL2 CCL3 CCL4 CCL11 CXCL1 CXCL10 and more
Catalog No. 50-000-81996 Supplier Meso Scale Discovery Supplier No. K152XCG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit is a multiplex immunoassay measuring 30 human cytokines and chemokines in serum plasma and culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and adhesive seals The assay runs in 3 5 hours quantifying IL-1 IL-2 IL-4 IL-5 IL-6 IL-7 IL-8 IL-10 IL-12p70 IL-13 IL-15 IL-16 IL-17A TNF- IFN- GM-CSF VEGF-A CCL2 CCL3 CCL4 CCL11 CXCL1 CXCL10 and more