Learn More
Meso Scale Discovery V-PLEX Plus Mouse IP-10 Kit
SDP
Catalog No. 5000080038
Encompass
The V-PLEX Plus Mouse IP-10 Kit from Meso Scale Discovery is an immunoassay that quantitatively measures Interferon gamma-induced protein 10 (IP-10) in mouse serum plasma and cell culture supernatants The kit contains a 96-well plate coated with IP-10 capture antibody detection antibody read buffer wash buffer blocker solutions calibrators and controls The electrochemiluminescence immunoassay has a sensitivity of 1 53 pg/mL and an assay range of 1 53-2500 pg/mL The total assay time is 3 5 hours This quantitative immunoassay kit allows for the accurate measurement of IP-10 levels in mouse biological samples
Catalog No. 50-000-80038 Supplier Meso Scale Discovery Supplier No. K152NVG1
The V-PLEX Plus Mouse IP-10 Kit from Meso Scale Discovery is an immunoassay that quantitatively measures Interferon gamma-induced protein 10 (IP-10) in mouse serum plasma and cell culture supernatants The kit contains a 96-well plate coated with IP-10 capture antibody detection antibody read buffer wash buffer blocker solutions calibrators and controls The electrochemiluminescence immunoassay has a sensitivity of 1 53 pg/mL and an assay range of 1 53-2500 pg/mL The total assay time is 3 5 hours This quantitative immunoassay kit allows for the accurate measurement of IP-10 levels in mouse biological samples