Learn More
Meso Scale Discovery V-PLEX Plus Mouse IP-10 Kit
SDP
Catalog No. 5000080040
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IP-10 Kit is an electrochemiluminescent immunoassay for quantifying IP-10 in mouse serum plasma and cell culture supernatants It includes a 96-well plate with IP-10 capture antibody detection antibody with an electrochemiluminescent label calibrator diluent wash buffer read buffer and plate seals The assay s sensitivity is 1 3 pg/mL with an upper quantification limit of 10 000 pg/mL It offers high sensitivity a wide dynamic range and rapid results testing up to 94 samples in duplicates with calibrator and controls
Catalog No. 50-000-80040 Supplier Meso Scale Discovery Supplier No. K152NVG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Mouse IP-10 Kit is an electrochemiluminescent immunoassay for quantifying IP-10 in mouse serum plasma and cell culture supernatants It includes a 96-well plate with IP-10 capture antibody detection antibody with an electrochemiluminescent label calibrator diluent wash buffer read buffer and plate seals The assay s sensitivity is 1 3 pg/mL with an upper quantification limit of 10 000 pg/mL It offers high sensitivity a wide dynamic range and rapid results testing up to 94 samples in duplicates with calibrator and controls