Learn More
Meso Scale Discovery V-PLEX Plus Mouse KC/GRO Kit
SDP
Catalog No. 5000078307
Encompass
The V-PLEX Plus Mouse KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay for measuring mouse keratinocyte chemoattractant (KC) and growth regulated oncogene (GRO) in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Diluent and lyophilized standards Using electrochemiluminescence it offers high sensitivity and a broad dynamic range with a small sample volume delivering reproducible results in about 5 hours
Catalog No. 50-000-78307 Supplier Meso Scale Discovery Supplier No. K152QTG1
The V-PLEX Plus Mouse KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay for measuring mouse keratinocyte chemoattractant (KC) and growth regulated oncogene (GRO) in serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Diluent and lyophilized standards Using electrochemiluminescence it offers high sensitivity and a broad dynamic range with a small sample volume delivering reproducible results in about 5 hours