Learn More
Meso Scale Discovery V-PLEX Plus Mouse MCP-1 Kit
SDP
Catalog No. 5000079252
Encompass
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit is a multiplex immunoassay measuring 30 cytokines and chemokines in serum plasma and culture supernatants It uses electrochemiluminescence for high sensitivity and dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and seals Sensitivity is 0 5-2 pg/mL requiring 50 L of sample Analytes include IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- GM-CSF VEGF-A MCP-1 Eotaxin IP-10 MIP-1 MIP-1 EGF FGF-2 HGF IFN- 2 MIG RANTES Eotaxin-3 TARC IL-5 IL-7 IL-15 IL-16 and IL-17A
Catalog No. 50-000-79252 Supplier Meso Scale Discovery Supplier No. K152NNG2
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit is a multiplex immunoassay measuring 30 cytokines and chemokines in serum plasma and culture supernatants It uses electrochemiluminescence for high sensitivity and dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and seals Sensitivity is 0 5-2 pg/mL requiring 50 L of sample Analytes include IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- GM-CSF VEGF-A MCP-1 Eotaxin IP-10 MIP-1 MIP-1 EGF FGF-2 HGF IFN- 2 MIG RANTES Eotaxin-3 TARC IL-5 IL-7 IL-15 IL-16 and IL-17A