Learn More
Meso Scale Discovery V-PLEX Plus Mouse MIP-1 Kit
SDP
Catalog No. 5000081381
Encompass
The Meso Scale Discovery V-PLEX Mouse MIP-1 alpha Kit is a multiplex immunoassay that quantitatively measures mouse MIP-1 alpha in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and assay diluent The kit has a detection range of 1 5-2500 pg/mL and requires 25 L of sample per well
Catalog No. 50-000-81381 Supplier Meso Scale Discovery Supplier No. K152NQG2
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX Mouse MIP-1 alpha Kit is a multiplex immunoassay that quantitatively measures mouse MIP-1 alpha in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and assay diluent The kit has a detection range of 1 5-2500 pg/mL and requires 25 L of sample per well