Learn More
Meso Scale Discovery V-PLEX Plus Mouse MIP-2 Kit
SDP
Catalog No. 5000078486
Encompass
The Meso Scale Discovery V-PLEX Mouse MIP-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse macrophage inflammatory protein 2 (MIP-2) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrator diluent wash buffer read buffer and vials of calibrator to generate a standard curve The assay has a detection range of 0 03-500 pg/mL and includes reagents for one 96-well plate This product can quantify mouse MIP-2 with high sensitivity and wide dynamic range
Catalog No. 50-000-78486 Supplier Meso Scale Discovery Supplier No. K152QCG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Mouse MIP-2 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse macrophage inflammatory protein 2 (MIP-2) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrator diluent wash buffer read buffer and vials of calibrator to generate a standard curve The assay has a detection range of 0 03-500 pg/mL and includes reagents for one 96-well plate This product can quantify mouse MIP-2 with high sensitivity and wide dynamic range