Learn More
Meso Scale Discovery V-PLEX Plus Mouse MIP-2 Kit
SDP
Catalog No. 5000082387
Encompass
The V-PLEX Plus Mouse MIP-2 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of mouse MIP-2 in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against mouse MIP-2 detection antibodies conjugated to an electrochemiluminescent label calibrator solutions diluent wash buffer read buffer and plate seals The assay has a detection range of 1 5-1000 pg/mL and exhibits minimal cross-reactivity with other cytokines The kit allows for high-sensitivity quantification of mouse MIP-2 levels from small sample volumes
Catalog No. 50-000-82387 Supplier Meso Scale Discovery Supplier No. K152QCG1
The V-PLEX Plus Mouse MIP-2 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of mouse MIP-2 in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against mouse MIP-2 detection antibodies conjugated to an electrochemiluminescent label calibrator solutions diluent wash buffer read buffer and plate seals The assay has a detection range of 1 5-1000 pg/mL and exhibits minimal cross-reactivity with other cytokines The kit allows for high-sensitivity quantification of mouse MIP-2 levels from small sample volumes