Learn More
Meso Scale Discovery V-PLEX Plus Mouse MIP-2 Kit
SDP
Catalog No. 5000082388
Encompass
Meso Scale Discovery offers a wide variety of V-PLEX assay kits for measuring biomarkers in mouse samples The electrochemiluminescent immunoassays provide high sensitivity broad dynamic range and low sample volume requirements Kits are available for measuring cytokines chemokines growth factors soluble receptors and other analytes in mouse serum plasma cell culture supernatants and other sample types The assays utilize MSD s proprietary MULTI-ARRAY technology and are compatible with their SECTOR Imager for rapid results With minimal hands-on time the V-PLEX kits allow efficient and accurate quantification of mouse biomarkers
Catalog No. 50-000-82388 Supplier Meso Scale Discovery Supplier No. K152QCG2
Meso Scale Discovery offers a wide variety of V-PLEX assay kits for measuring biomarkers in mouse samples The electrochemiluminescent immunoassays provide high sensitivity broad dynamic range and low sample volume requirements Kits are available for measuring cytokines chemokines growth factors soluble receptors and other analytes in mouse serum plasma cell culture supernatants and other sample types The assays utilize MSD s proprietary MULTI-ARRAY technology and are compatible with their SECTOR Imager for rapid results With minimal hands-on time the V-PLEX kits allow efficient and accurate quantification of mouse biomarkers