Learn More
Meso Scale Discovery V-PLEX Plus Neuroinflammation Panel 1 Human Kit, Multiplex Immunoassay, 96-well plate, 37 biomarkers, Detection limits: sub- to low pg/mL, Research use only
SDP
Catalog No. NC3666910
Encompass
The V-PLEX Plus Neuroinflammation Panel 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay designed for the quantitative measurement of 37 human biomarkers associated with neuroinflammatory responses. It employs electrochemiluminescence (ECL) MULTI-ARRAY technology to ensure accurate and reproducible results. The kit is validated for various sample types including serum, plasma, and cerebrospinal fluid, and is intended for research use only.
  • 37 human biomarkers measured
  • High sensitivity with detection limits in sub- to low pg/mL range
  • Wide linear dynamic range
  • Lot-to-lot consistency
  • Validated for multiple sample types
  • Controls, wash buffer, and plate seals included
  • 96-well plate type
  • Research use only
Catalog No. NC3666910 Supplier Meso Scale Discovery Supplier No. K15210G2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Neuroinflammation Panel 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay designed for the quantitative measurement of 37 human biomarkers associated with neuroinflammatory responses. It employs electrochemiluminescence (ECL) MULTI-ARRAY technology to ensure accurate and reproducible results. The kit is validated for various sample types including serum, plasma, and cerebrospinal fluid, and is intended for research use only.
  • 37 human biomarkers measured
  • High sensitivity with detection limits in sub- to low pg/mL range
  • Wide linear dynamic range
  • Lot-to-lot consistency
  • Validated for multiple sample types
  • Controls, wash buffer, and plate seals included
  • 96-well plate type
  • Research use only