Learn More
Meso Scale Discovery V-PLEX Plus NHP Eotaxin-3 Kit
SDP
Catalog No. 5000080776
Encompass
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit is a multiplex immunoassay that measures 30 human cytokines and chemokines in a single small sample volume It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators controls diluent read buffer and wash buffer It can be used to profile cytokine expression and immune responses The kit has been optimized for serum plasma culture supernatants and other sample types
Catalog No. 50-000-80776 Supplier Meso Scale Discovery Supplier No. K156NUG2
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit is a multiplex immunoassay that measures 30 human cytokines and chemokines in a single small sample volume It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators controls diluent read buffer and wash buffer It can be used to profile cytokine expression and immune responses The kit has been optimized for serum plasma culture supernatants and other sample types