Learn More
Meso Scale Discovery V-PLEX Plus NHP Eotaxin-3 Kit
SDP
Catalog No. 5000081280
Encompass
The Meso Scale Discovery V-PLEX Plus NHP Eotaxin-3 Kit is a sensitive immunoassay for measuring Eotaxin-3 in non-human primate serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity and a wide dynamic range requiring only 50 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and diluents for rhesus and cynomolgus samples enabling accurate Eotaxin-3 quantitation for immunology and inflammation research
Catalog No. 50-000-81280 Supplier Meso Scale Discovery Supplier No. K156NUG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus NHP Eotaxin-3 Kit is a sensitive immunoassay for measuring Eotaxin-3 in non-human primate serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity and a wide dynamic range requiring only 50 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and diluents for rhesus and cynomolgus samples enabling accurate Eotaxin-3 quantitation for immunology and inflammation research