Learn More
Meso Scale Discovery V-PLEX Plus NHP GM-CSF Kit
SDP
Catalog No. 5000080823
Encompass
The Meso Scale Discovery MULTI-ARRAY GM-CSF Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human Granulocyte-Macrophage Colony Stimulating Factor (GM-CSF) in serum plasma and cell culture supernatants This assay utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL The assay requires 25 L of sample and can be read on MSD s SECTOR Imager instruments This assay allows for the rapid accurate and reliable measurement of GM-CSF levels
Catalog No. 50-000-80823 Supplier Meso Scale Discovery Supplier No. K156RIG2
The Meso Scale Discovery MULTI-ARRAY GM-CSF Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human Granulocyte-Macrophage Colony Stimulating Factor (GM-CSF) in serum plasma and cell culture supernatants This assay utilizes MSD s patented SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The assay has a lower limit of detection of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL The assay requires 25 L of sample and can be read on MSD s SECTOR Imager instruments This assay allows for the rapid accurate and reliable measurement of GM-CSF levels