Learn More
Meso Scale Discovery V-PLEX Plus NHP GM-CSF Kit
SDP
Catalog No. 5000081244
Encompass
The Meso Scale Discovery MULTI-ARRAY GM-CSF Assay is an electrochemiluminescent immunoassay for quantifying GM-CSF in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results in 3 5 hours The detection limit is 0 04 pg/mL and the quantification limit is 1500 pg/mL It requires 50 L of sample and shows no cross-reactivity with other cytokines The kit includes a 96-well plate detection antibody buffers calibrators and controls For research use only
Catalog No. 50-000-81244 Supplier Meso Scale Discovery Supplier No. K156RIG1
The Meso Scale Discovery MULTI-ARRAY GM-CSF Assay is an electrochemiluminescent immunoassay for quantifying GM-CSF in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results in 3 5 hours The detection limit is 0 04 pg/mL and the quantification limit is 1500 pg/mL It requires 50 L of sample and shows no cross-reactivity with other cytokines The kit includes a 96-well plate detection antibody buffers calibrators and controls For research use only