Learn More
Meso Scale Discovery V-PLEX Plus NHP GM-CSF Kit
SDP
Catalog No. 5000081550
Encompass
The Meso Scale Discovery MULTI-ARRAY Human GM-CSF Kit is a 4-plate assay kit for the quantitative measurement of human GM-CSF in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity wide dynamic range and low sample volume requirements This kit includes plates coated with capture antibody detection antibody read buffer sample diluent calibrator and control It can detect GM-CSF concentrations from 0 2-2000 pg/mL with an assay time of 3 5 hours
Catalog No. 50-000-81550 Supplier Meso Scale Discovery Supplier No. K156RIG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery MULTI-ARRAY Human GM-CSF Kit is a 4-plate assay kit for the quantitative measurement of human GM-CSF in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity wide dynamic range and low sample volume requirements This kit includes plates coated with capture antibody detection antibody read buffer sample diluent calibrator and control It can detect GM-CSF concentrations from 0 2-2000 pg/mL with an assay time of 3 5 hours