Learn More
Meso Scale Discovery V-PLEX Plus NHP IFN-y Kit
SDP
Catalog No. 5000079063
Encompass
The V-PLEX Plus NHP IFN gamma Kit from Meso Scale Discovery quantifies interferon gamma (IFN-) in non-human primate samples like serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies and calibrators for a standard curve It detects IFN- from 0 13 pg/mL to 10 000 pg/mL in 2 hours aiding immunology and inflammation research
Catalog No. 50-000-79063 Supplier Meso Scale Discovery Supplier No. K156QOG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus NHP IFN gamma Kit from Meso Scale Discovery quantifies interferon gamma (IFN-) in non-human primate samples like serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies ECL-labeled detection antibodies and calibrators for a standard curve It detects IFN- from 0 13 pg/mL to 10 000 pg/mL in 2 hours aiding immunology and inflammation research