Learn More
Meso Scale Discovery V-PLEX Plus NHP IFN-y Kit
SDP
Catalog No. 5000080802
Encompass
The V-PLEX Plus NHP IFN gamma Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay kit for the quantitative measurement of interferon gamma (IFN-) in non-human primate (NHP) samples such as serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrator diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 49 pg/mL and a range from 0 49-2500 pg/mL It uses a sandwich immunoassay format and takes approximately 3 5 hours to run The kit has high sensitivity wide dynamic range and requires low sample volume
Catalog No. 50-000-80802 Supplier Meso Scale Discovery Supplier No. K156QOG1
The V-PLEX Plus NHP IFN gamma Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay kit for the quantitative measurement of interferon gamma (IFN-) in non-human primate (NHP) samples such as serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound calibrator diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 49 pg/mL and a range from 0 49-2500 pg/mL It uses a sandwich immunoassay format and takes approximately 3 5 hours to run The kit has high sensitivity wide dynamic range and requires low sample volume