Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-10 Kit
SDP
Catalog No. 5000080803
Encompass
The V-PLEX Plus NHP IL-10 Kit from Meso Scale Discovery is a 96-well plate assay using electrochemiluminescence (ECL) for quantifying natural human IL-10 in various biological fluids It offers high sensitivity a wide dynamic range and low sample volume requirements The kit includes a plate with capture antibody ECL-labeled detection antibody calibrators diluent wash buffer read buffer and plate seals The assay takes 3 5 hours with a 2-hour incubation and detects 1 53-1500 pg/mL It is ideal for measuring natural human IL-10
Catalog No. 50-000-80803 Supplier Meso Scale Discovery Supplier No. K156QUG1
The V-PLEX Plus NHP IL-10 Kit from Meso Scale Discovery is a 96-well plate assay using electrochemiluminescence (ECL) for quantifying natural human IL-10 in various biological fluids It offers high sensitivity a wide dynamic range and low sample volume requirements The kit includes a plate with capture antibody ECL-labeled detection antibody calibrators diluent wash buffer read buffer and plate seals The assay takes 3 5 hours with a 2-hour incubation and detects 1 53-1500 pg/mL It is ideal for measuring natural human IL-10