Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-10 Kit
SDP
Catalog No. 5000080828
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-10 Kit is a 5-plate assay kit for the quantitative measurement of non-human primate interleukin 10 (IL-10) in serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes plates coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-80828 Supplier Meso Scale Discovery Supplier No. K156QUG2
The Meso Scale Discovery V-PLEX Plus NHP IL-10 Kit is a 5-plate assay kit for the quantitative measurement of non-human primate interleukin 10 (IL-10) in serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes plates coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL