Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-15 Kit
SDP
Catalog No. 5000078991
Encompass
The V-PLEX Plus NHP IL-15 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of natural human IL-15 in serum plasma and cell culture supernatants The kit includes a plate coated with a monoclonal antibody specific for human IL-15 a ruthenium-labeled detection antibody and calibrators and controls to generate a standard curve and validate assay performance The sensitive chemiluminescent signal provides a dynamic range from 3 2 to 10 000 pg/mL with high specificity and no cross-reactivity to other cytokines This IL-15 assay kit offers a rapid 3 5 hour protocol in a convenient 96-well format
Catalog No. 50-000-78991 Supplier Meso Scale Discovery Supplier No. K156RDG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus NHP IL-15 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of natural human IL-15 in serum plasma and cell culture supernatants The kit includes a plate coated with a monoclonal antibody specific for human IL-15 a ruthenium-labeled detection antibody and calibrators and controls to generate a standard curve and validate assay performance The sensitive chemiluminescent signal provides a dynamic range from 3 2 to 10 000 pg/mL with high specificity and no cross-reactivity to other cytokines This IL-15 assay kit offers a rapid 3 5 hour protocol in a convenient 96-well format