Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-15 Kit
SDP
Catalog No. 5000079938
Encompass
The K156RDG-1 is a thermoelectric module from Mesoscale that provides 156 Watts of cooling power at a maximum hot side temperature of 30 C and a 15 C temperature differential It uses Bismuth Telluride semiconductor material and incorporates a ceramic substrate and thermally conductive alumina insulators The module has a square shape with dimensions of 40 x 40 x 3 4mm It requires a cold side heat sink and fan for proper operation With a no load voltage of 3 7V and operating current of 15 Amps this Peltier cooler is well-suited for applications requiring high heat pumping capacity in a compact footprint
Catalog No. 50-000-79938 Supplier Meso Scale Discovery Supplier No. K156RDG1
The K156RDG-1 is a thermoelectric module from Mesoscale that provides 156 Watts of cooling power at a maximum hot side temperature of 30 C and a 15 C temperature differential It uses Bismuth Telluride semiconductor material and incorporates a ceramic substrate and thermally conductive alumina insulators The module has a square shape with dimensions of 40 x 40 x 3 4mm It requires a cold side heat sink and fan for proper operation With a no load voltage of 3 7V and operating current of 15 Amps this Peltier cooler is well-suited for applications requiring high heat pumping capacity in a compact footprint