Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-16 Kit
SDP
Catalog No. 5000079917
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-16 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 16 (IL-16) in serum plasma and cell culture supernatants The kit contains a plate coated with a monoclonal antibody to human IL-16 a ruthenium-labeled detection antibody calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a detection range of 0 06-2500 pg/mL and takes approximately 3 5 hours to complete This product can aid in understanding immune system function and inflammation
Catalog No. 50-000-79917 Supplier Meso Scale Discovery Supplier No. K156REG2
The Meso Scale Discovery V-PLEX Plus NHP IL-16 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 16 (IL-16) in serum plasma and cell culture supernatants The kit contains a plate coated with a monoclonal antibody to human IL-16 a ruthenium-labeled detection antibody calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a detection range of 0 06-2500 pg/mL and takes approximately 3 5 hours to complete This product can aid in understanding immune system function and inflammation