Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-17A Kit
SDP
Catalog No. 5000079936
Encompass
The V-PLEX Plus NHP IL-17A Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-17A in non-human primate serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range and shows no cross-reactivity with other cytokines The kit includes a 96-well plate coated with IL-17A capture antibody detection antibody read buffer wash buffer diluent and calibrators and controls The total assay time is 3 5 hours This kit allows reliable reproducible quantification of IL-17A from NHP samples
Catalog No. 50-000-79936 Supplier Meso Scale Discovery Supplier No. K156RFG2
The V-PLEX Plus NHP IL-17A Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-17A in non-human primate serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range and shows no cross-reactivity with other cytokines The kit includes a 96-well plate coated with IL-17A capture antibody detection antibody read buffer wash buffer diluent and calibrators and controls The total assay time is 3 5 hours This kit allows reliable reproducible quantification of IL-17A from NHP samples