Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-1SY Kit
SDP
Catalog No. 5000079064
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-1 in non-human primate serum plasma and culture supernatants The kit contains a plate coated with a capture antibody a detection antibody conjugated to an electrochemiluminescent label and calibrators and controls The assay has a sensitivity of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL Results are obtained in as little as 3 5 hours using an MSD instrument capable of reading electrochemiluminescence This assay allows for the accurate quantification of IL-1 levels in non-human primate samples
Catalog No. 50-000-79064 Supplier Meso Scale Discovery Supplier No. K156QPG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus NHP IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-1 in non-human primate serum plasma and culture supernatants The kit contains a plate coated with a capture antibody a detection antibody conjugated to an electrochemiluminescent label and calibrators and controls The assay has a sensitivity of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL Results are obtained in as little as 3 5 hours using an MSD instrument capable of reading electrochemiluminescence This assay allows for the accurate quantification of IL-1 levels in non-human primate samples