Learn More
Meso Scale Discovery V-PLEX Plus NHP IL-1SY Kit
SDP
Catalog No. 5000081142
Encompass
The V-PLEX Plus NHP IL-1 Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) for measuring interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and other samples It includes kits for both human and non-human primate IL-1 detection The assay is highly sensitive detecting IL-1 as low as 0 06 pg/mL The kit features a 96-well plate with IL-1 capture antibody detection antibody buffers and a calibration curve from 3 2 - 10 000 pg/mL aiding research on inflammation and immune response
Catalog No. 50-000-81142 Supplier Meso Scale Discovery Supplier No. K156QPG2
The V-PLEX Plus NHP IL-1 Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) for measuring interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and other samples It includes kits for both human and non-human primate IL-1 detection The assay is highly sensitive detecting IL-1 as low as 0 06 pg/mL The kit features a 96-well plate with IL-1 capture antibody detection antibody buffers and a calibration curve from 3 2 - 10 000 pg/mL aiding research on inflammation and immune response